Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042655.1 | NP_001036120.1 | 8 - 9 | 1138 | 78 | |
GenBank mRNA | AK039566.1 | - | 8 - 9 | 953 | 78 | |
AK046883.1 | - | 8 - 9 | 972 | 78 | ||
AK075571.1 | - | 2 - 3 | 242 | 78 | ||
AK081318.1 | - | 8 - 9 | 951 | 78 | ||
AK083153.1 | - | 8 - 9 | 956 | 78 | ||
AK087754.1 | - | 963 | 78 | |||
AK136705.1 | - | 8 - 9 | 963 | 78 | ||
AK149917.1 | - | 8 - 9 | 950 | 78 | ||
AK153732.1 | - | 8 - 9 | 950 | 78 | ||
AK170821.1 | - | 8 - 9 | 950 | 78 | ||
BC017607.1 | - | 8 - 9 | 1138 | 4507589 | 78 | |
BC035449.1 | - | 8 - 9 | 937 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |