Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285997.1 | NP_001272926.1 | 4 - 5 | 633 | 147 | |
NM_001285998.1 | NP_001272927.1 | 4 - 5 | 633 | 147 | ||
NM_145150.3 | NP_660132.2 | 4 - 5 | 633 | 147 | ||
GenBank mRNA | AK029140.1 | - | 4 - 5 | 620 | 147 | |
AK088576.1 | - | 4 - 5 | 605 | 147 | ||
AK135778.1 | - | 2 - 3 | 258 | 147 | ||
BC005475.1 | - | 4 - 5 | 567 | 3592138 | 147 | |
BC059808.1 | - | 4 - 5 | 578 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |