Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145519.2 | NP_663494.2 | 6 - 7 | 619 | 68 | |
GenBank mRNA | AK050860.1 | - | 6 - 7 | 579 | 68 | |
AK086699.1 | - | 6 - 7 | 613 | 68 | ||
AK171713.1 | - | 6 - 7 | 645 | 68 | ||
BC009153.1 | - | 6 - 7 | 608 | 2655209 | 68 | |
BI739824.1 | - | 4 - 5 | 244 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |