Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083312.2 | NP_001076781.1 | 1 - 2 | 107 | 66 | |
GenBank mRNA | AK087751.1 | - | 1 - 2 | 101 | 66 | |
BC010229.1 | - | 1 - 2 | 72 | 3501441 | 66 | |
BY742706.1 | - | 1 - 2 | 105 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |