Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145916.2 | NP_666021.2 | 1 - 2 | 84 | 67 | |
GenBank mRNA | BC011501.1 | - | 1 - 2 | 102 | 3584546 | 67 |
BU613201.1 | - | 1 - 2 | 84 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |