Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145949.2 | NP_666061.3 | 6 - 7 | 636 | 87 | |
GenBank mRNA | AK082385.1 | - | 5 - 6 | 827 | 87 | |
BC026393.1 | - | 6 - 7 | 636 | 4195134 | 87 | |
EF137182.1 | - | 6 - 7 | 636 | 87 | ||
EU440733.1 | - | 5 - 6 | 428 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |