Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145970.1 | NP_666082.2 | 10 - 11 | 1182 | 67 | |
GenBank mRNA | BC016188.1 | - | 10 - 11 | 1182 | 4482793 | 67 |
BC027028.1 | - | 9 - 10 | 997 | 5363547 | 67 | |
DQ329239.1 | - | 3 - 4 | 232 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |