Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145978.2 | NP_666090.1 | 1 - 2 | 62 | 65 | |
GenBank mRNA | BC024556.1 | - | 1 - 2 | 62 | 4990983 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |