Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163622.1 | NP_001157094.1 | 8 - 9 | 1640 | 89 | |
NM_001163623.1 | NP_001157095.1 | 7 - 8 | 1104 | 89 | ||
NM_001163624.1 | NP_001157096.1 | 7 - 8 | 972 | 89 | ||
NM_145984.3 | NP_666096.3 | 8 - 9 | 1204 | 89 | ||
GenBank mRNA | AK028877.1 | - | 8 - 9 | 1598 | 89 | |
AK050878.1 | - | 7 - 8 | 966 | 89 | ||
AK083070.1 | - | 8 - 9 | 1145 | 89 | ||
AK129142.1 | - | 2 - 3 | 1058 | 89 | ||
BC004612.1 | - | 7 - 8 | 1058 | 3585402 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |