Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145987.2 | NP_666099.2 | 2 - 3 | 808 | 84 | |
GenBank mRNA | AK085940.1 | - | 808 | 84 | ||
AK154809.1 | - | 2 - 3 | 194 | 84 | ||
AK162322.1 | - | 2 - 3 | 157 | 84 | ||
BC025833.1 | - | 2 - 3 | 209 | 5125918 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |