Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289711.1 | NP_001276640.1 | 3 - 4 | 510 | 78 | |
NM_001289712.1 | NP_001276641.1 | 3 - 4 | 510 | 78 | ||
NM_145993.5 | NP_666105.2 | 3 - 4 | 510 | 78 | ||
NR_110362.1 | 3 - 4 | 510 | 78 | |||
GenBank mRNA | AK029115.1 | - | 3 - 4 | 455 | 78 | |
AK036510.1 | - | 3 - 4 | 463 | 78 | ||
AK132012.1 | - | 3 - 4 | 463 | 78 | ||
AK141810.1 | - | 2 - 3 | 1644 | 78 | ||
AK145560.1 | - | 3 - 4 | 452 | 78 | ||
AK146632.1 | - | 3 - 4 | 466 | 78 | ||
AK156236.1 | - | 3 - 4 | 465 | 78 | ||
AY237000.1 | - | 3 - 4 | 443 | 78 | ||
AY237001.1 | - | 3 - 4 | 443 | 78 | ||
AY237002.1 | - | 3 - 4 | 443 | 78 | ||
BC023933.1 | - | 3 - 4 | 451 | 5325131 | 78 | |
BC030864.1 | - | 3 - 4 | 413 | 4211489 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |