Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 10 - 11 | 1820 | 105 | |
GenBank mRNA | AK144877.1 | - | 10 - 11 | 1791 | 105 | |
AK159079.1 | - | 10 - 11 | 1797 | 105 | ||
AK166055.1 | - | 10 - 11 | 1801 | 105 | ||
BC010717.1 | - | 10 - 11 | 1461 | 105 | ||
BC011506.3 | - | 10 - 11 | 1754 | 105 | ||
BC080691.1 | - | 10 - 11 | 1506 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |