Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177798.1 | NP_001171269.1 | 9 - 10 | 1168 | 63 | |
NM_146092.2 | NP_666204.2 | 9 - 10 | 1005 | 63 | ||
GenBank mRNA | AK077149.1 | - | 9 - 10 | 1169 | 63 | |
AK209124.1 | - | 1 - 2 | 76 | 63 | ||
BC020299.1 | - | 9 - 10 | 978 | 63 | ||
BC028647.1 | - | 9 - 10 | 952 | 1244243 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |