Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146097.3 | NP_666209.1 | 3 - 4 | 417 | 116 | |
NR_033744.1 | 3 - 4 | 484 | 116 | |||
GenBank mRNA | AK076027.1 | - | 3 - 4 | 372 | 116 | |
AK132291.1 | - | 3 - 4 | 403 | 116 | ||
BC018472.1 | - | 3 - 4 | 359 | 3661842 | 116 | |
BC053745.1 | - | 3 - 4 | 452 | 6505520 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |