Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146136.1 | NP_666248.1 | 1 - 2 | 125 | 130 | |
GenBank mRNA | AK045797.1 | - | 1 - 2 | 111 | 130 | |
AK049873.1 | - | 1 - 2 | 188 | 130 | ||
AK052779.1 | - | 1 - 2 | 124 | 130 | ||
AK085492.1 | - | 1 - 2 | 122 | 130 | ||
AK136837.1 | - | 1 - 2 | 124 | 130 | ||
BC025441.1 | - | 1 - 2 | 113 | 5043709 | 130 | |
BC026596.1 | - | 1 - 2 | 125 | 4975010 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |