Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146176.3 | NP_666288.1 | 1 - 2 | 315 | 104 | |
GenBank mRNA | AK141718.1 | - | 1 - 2 | 295 | 104 | |
BC030332.1 | - | 1 - 2 | 244 | 5400871 | 104 | |
BC053437.1 | - | 1 - 2 | 76 | 6394085 | 104 | |
BC141041.1 | - | 1 - 2 | 115 | 9055653 | 104 | |
CV783268.1 | - | 1 - 2 | 317 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |