Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036740.2 | NP_001031817.1 | 1 - 2 | 98 | 83 | |
GenBank mRNA | AK154901.1 | - | 1 - 2 | 54 | 83 | |
BC025206.1 | - | 1 - 2 | 84 | 5037295 | 83 | |
BY320287.1 | - | 1 - 2 | 82 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |