Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146188.1 | NP_666300.1 | 4 - 5 | 672 | 75 | |
GenBank mRNA | AK034731.1 | - | 5 - 6 | 744 | 75 | |
BC031749.1 | - | 4 - 5 | 672 | 4507203 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |