Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146219.1 | NP_666331.1 | 3 - 4 | 1294 | 117 | |
GenBank mRNA | AK131135.1 | - | 3 - 4 | 1219 | 117 | |
AK157027.1 | - | 3 - 4 | 1328 | 117 | ||
AK164831.1 | - | 3 - 4 | 1329 | 117 | ||
BC025816.1 | - | 3 - 4 | 1294 | 5098064 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |