Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110253.2 | NP_001103723.2 | 16 - 17 | 4341 | 76 | |
NM_148925.3 | NP_683727.3 | 17 - 18 | 4588 | 76 | ||
GenBank mRNA | AJ428065.1 | - | 17 - 18 | 4493 | 76 | |
AK044587.1 | - | 9 - 10 | 1650 | 76 | ||
BC022708.1 | - | 4 - 5 | 593 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |