Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009009.4 | NP_033035.3 | 2 - 3 | 395 | 78 | |
GenBank mRNA | AF332085.1 | - | 1 - 2 | 150 | 78 | |
AF332086.1 | - | 1 - 2 | 150 | 78 | ||
AK004746.1 | - | 2 - 3 | 198 | 78 | ||
AK128984.1 | - | 2 - 3 | 284 | 78 | ||
AK139492.1 | - | 1 - 2 | 48 | 78 | ||
AK145591.1 | - | 2 - 3 | 362 | 78 | ||
AK163992.1 | - | 256 | 78 | |||
AK168718.1 | - | 2 - 3 | 383 | 78 | ||
AK168896.1 | - | 2 - 3 | 344 | 78 | ||
BC043032.1 | - | 1 - 2 | 164 | 5696540 | 78 | |
BC129916.1 | - | 1 - 2 | 194 | 40111820 | 78 | |
BC129917.1 | - | 1 - 2 | 195 | 40111821 | 78 | |
D37790.1 | - | 1 - 2 | 82 | 78 | ||
X98293.1 | - | 1 - 2 | 150 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |