Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015786.3 | NP_056601.1 | 1 | 60 | 81 | |
GenBank mRNA | AK002321.1 | - | 62 | 81 | ||
AK155502.1 | - | 61 | 81 | |||
BC089604.1 | - | 60 | 6763721 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |