Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089086.1 | - | 10 - 11 | 959 | 68 | |
AK089169.1 | - | 10 - 11 | 981 | 68 | ||
AK134118.1 | - | 1 - 2 | 222 | 68 | ||
AK150411.1 | - | 10 - 11 | 961 | 68 | ||
AK151535.1 | - | 2 - 3 | 190 | 68 | ||
AK152675.1 | - | 10 - 11 | 978 | 68 | ||
AK152769.1 | - | 10 - 11 | 960 | 68 | ||
AK153065.1 | - | 10 - 11 | 960 | 68 | ||
AK155772.1 | - | 10 - 11 | 959 | 68 | ||
AK155912.1 | - | 10 - 11 | 979 | 68 | ||
AK160041.1 | - | 10 - 11 | 962 | 68 | ||
AK170019.1 | - | 10 - 11 | 984 | 68 | ||
AK171022.1 | - | 10 - 11 | 981 | 68 | ||
AK171060.1 | - | 10 - 11 | 959 | 68 | ||
BC025584.1 | - | 10 - 11 | 1013 | 5324943 | 68 | |
X13335.1 | - | 10 - 11 | 958 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |