Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037723.3 | NP_001032812.2 | 21 - 22 | 2934 | 73 | |
NM_001037724.4 | NP_001032813.1 | 21 - 22 | 2870 | 73 | ||
NM_001109756.1 | NP_001103226.1 | 21 - 22 | 2838 | 73 | ||
NM_007406.2 | NP_031432.2 | 21 - 22 | 3022 | 73 | ||
GenBank mRNA | AK138911.1 | - | 21 - 22 | 2854 | 73 | |
AK154262.1 | - | 21 - 22 | 2925 | 73 | ||
AK154652.1 | - | 21 - 22 | 2840 | 73 | ||
AK155646.1 | - | 21 - 22 | 2839 | 73 | ||
AK155831.1 | - | 21 - 22 | 2841 | 73 | ||
BC115833.1 | - | 19 - 20 | 2716 | 40086640 | 73 | |
U12919.1 | - | 21 - 22 | 3023 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |