Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007434.3 | NP_031460.1 | 1 - 2 | 84 | 74 | |
GenBank mRNA | BC026151.1 | - | 1 - 2 | 107 | 4187425 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |