Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_001243043.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_001243043.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_001243044.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_001243044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_007454.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
NM_007454.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000667.mm.1 |
TC1100000058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_001243043.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_001243043.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_001243044.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_001243044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_007454.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
NM_007454.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000666.mm.1 |
TC1100000058.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_001243043.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_001243043.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_001243044.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_001243044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_007454.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
NM_007454.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000665.mm.1 |
TC1100000058.mm.1 |
MTA TC |