Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007462.3 | NP_031488.2 | 11 - 12 | 1715 | 106 | |
GenBank mRNA | AK046280.1 | - | 11 - 12 | 1705 | 106 | |
AK080907.1 | - | 11 - 12 | 1715 | 106 | ||
BC141141.1 | - | 10 - 11 | 1428 | 9055753 | 106 | |
M88127.1 | - | 10 - 11 | 1404 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |