Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009963.4 | NP_034093.1 | 9 - 10 | 1590 | 62 | |
GenBank mRNA | AB003433.1 | - | 9 - 10 | 1475 | 62 | |
AF156987.1 | - | 9 - 10 | 1558 | 62 | ||
AK041696.1 | - | 9 - 10 | 1558 | 62 | ||
AK133781.1 | - | 9 - 10 | 1565 | 62 | ||
AK172994.1 | - | 9 - 10 | 1522 | 62 | ||
BC006077.1 | - | 3 - 4 | 498 | 62 | ||
BC054794.1 | - | 9 - 10 | 1546 | 6834104 | 62 | |
BC066799.1 | - | 9 - 10 | 1549 | 30103169 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |