Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026083.2 | NP_080359.2 | 8 - 9 | 1241 | 103 | |
GenBank mRNA | AK014200.1 | - | 8 - 9 | 1021 | 103 | |
AK078225.1 | - | 8 - 9 | 1232 | 103 | ||
BC038882.1 | - | 1 - 2 | 51 | 103 | ||
BC138264.1 | - | 8 - 9 | 1190 | 8861283 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |