Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026792.3 | NP_081068.1 | 1 - 2 | 495 | 113 | |
GenBank mRNA | AK003649.1 | - | 1 - 2 | 239 | 113 | |
AK082137.1 | - | 1 - 2 | 270 | 113 | ||
AK089885.1 | - | 1 - 2 | 298 | 113 | ||
AK152709.1 | - | 1 - 2 | 238 | 113 | ||
AK152889.1 | - | 1 - 2 | 242 | 113 | ||
AK155378.1 | - | 2 - 3 | 444 | 113 | ||
AK170093.1 | - | 1 - 2 | 239 | 113 | ||
AY161042.1 | - | 1 - 2 | 472 | 113 | ||
BC031987.1 | - | 1 - 2 | 220 | 3982362 | 113 | |
BC062903.1 | - | 1 - 2 | 495 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |