Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028710.3 | NP_082986.3 | 5 - 6 | 1273 | 82 | |
GenBank mRNA | AK018132.2 | - | 2 - 3 | 690 | 82 | |
AK158726.1 | - | 5 - 6 | 1274 | 82 | ||
AK173082.1 | - | 2 - 3 | 372 | 82 | ||
BC022158.1 | - | 1 - 2 | 103 | 82 | ||
BC084731.1 | - | 2 - 3 | 963 | 6813261 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |