Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166177.1 | NP_001159649.1 | 2 - 3 | 222 | 108 | |
NM_028710.3 | NP_082986.3 | 11 - 12 | 1949 | 108 | ||
GenBank mRNA | AK018132.2 | - | 8 - 9 | 1366 | 108 | |
AK158726.1 | - | 11 - 12 | 1950 | 108 | ||
AK173082.1 | - | 8 - 9 | 1048 | 108 | ||
BC022158.1 | - | 7 - 8 | 779 | 108 | ||
BC039629.1 | - | 2 - 3 | 179 | 108 | ||
BC084731.1 | - | 8 - 9 | 1639 | 6813261 | 108 | |
BY095625.1 | - | 2 - 3 | 204 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |