Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028833.3 | NP_083109.2 | 12 - 13 | 1154 | 77 | |
GenBank mRNA | AK006472.2 | - | 11 - 12 | 895 | 77 | |
BC059223.1 | - | 12 - 13 | 1154 | 6417270 | 77 | |
BC065128.1 | - | 10 - 11 | 835 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |