Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164609.1 | NP_001158081.1 | 6 - 7 | 579 | 101 | |
NM_001164610.1 | NP_001158082.1 | 6 - 7 | 579 | 101 | ||
NM_001164611.1 | NP_001158083.1 | 6 - 7 | 579 | 101 | ||
NM_029945.3 | NP_084221.2 | 6 - 7 | 579 | 101 | ||
GenBank mRNA | AK019466.2 | - | 6 - 7 | 580 | 101 | |
AK031029.1 | - | 6 - 7 | 573 | 101 | ||
AK031685.1 | - | 6 - 7 | 578 | 101 | ||
AK035892.1 | - | 6 - 7 | 571 | 101 | ||
AK036055.1 | - | 6 - 7 | 571 | 101 | ||
AK036380.1 | - | 6 - 7 | 575 | 101 | ||
AK129354.1 | - | 6 - 7 | 556 | 101 | ||
AK138720.1 | - | 6 - 7 | 553 | 101 | ||
AK164234.1 | - | 6 - 7 | 571 | 101 | ||
AK164456.1 | - | 4 - 5 | 1044 | 101 | ||
BC026767.1 | - | 6 - 7 | 552 | 5137008 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |