Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172116.4 | NP_742114.3 | 5 - 6 | 517 | 62 | |
GenBank mRNA | AK081187.1 | - | 5 - 6 | 477 | 62 | |
AK084273.1 | - | 5 - 6 | 473 | 62 | ||
AK088159.1 | - | 5 - 6 | 493 | 62 | ||
AK140975.1 | - | 5 - 6 | 468 | 62 | ||
AK150323.1 | - | 3 - 4 | 1187 | 62 | ||
AK150354.1 | - | 3 - 4 | 1187 | 62 | ||
AK151259.1 | - | 3 - 4 | 1187 | 62 | ||
AK151794.1 | - | 3 - 4 | 1187 | 62 | ||
AK151959.1 | - | 3 - 4 | 1187 | 62 | ||
AK153110.1 | - | 3 - 4 | 1187 | 62 | ||
AK161205.1 | - | 4 - 5 | 1320 | 62 | ||
BC023835.1 | - | 5 - 6 | 487 | 5345186 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |