Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200017531.mm.1 |
NM_172117.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017531.mm.1 |
NM_172117.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017531.mm.1 |
TC0200002227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017530.mm.1 |
NM_172117.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017530.mm.1 |
NM_172117.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017530.mm.1 |
TC0200002227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017528.mm.1 |
NM_172117.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017528.mm.1 |
NM_172117.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017528.mm.1 |
TC0200002227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017532.mm.1 |
NM_172117.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017532.mm.1 |
NM_172117.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017532.mm.1 |
TC0200002227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017527.mm.1 |
NM_172117.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017527.mm.1 |
NM_172117.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017527.mm.1 |
TC0200002227.mm.1 |
MTA TC |