Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crocc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145958.1 | NP_001139430.1 | 34 - 35 | 5594 | 54 | |
NM_172122.2 | NP_742120.2 | 35 - 36 | 5847 | 54 | ||
GenBank mRNA | AF527975.1 | - | 35 - 36 | 5847 | 54 | |
AK032335.1 | - | 6 - 7 | 1510 | 54 | ||
AK122285.1 | - | 26 - 27 | 5090 | 54 | ||
BC027090.1 | - | 8 - 9 | 1388 | 54 | ||
BC034536.1 | - | 12 - 13 | 2752 | 54 | ||
BC036729.1 | - | 20 - 21 | 3489 | 54 | ||
BC038074.1 | - | 4 - 5 | 729 | 54 | ||
BC054054.1 | - | 33 - 34 | 5575 | 6394697 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |