Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172132.2 | NP_742144.1 | 19 - 20 | 2905 | 69 | |
GenBank mRNA | AK028962.1 | - | 8 - 9 | 1073 | 69 | |
AK043678.1 | - | 5 - 6 | 566 | 69 | ||
AK078074.1 | - | 19 - 20 | 2907 | 69 | ||
AK129233.1 | - | 19 - 20 | 2843 | 69 | ||
AK141879.1 | - | 19 - 20 | 2882 | 69 | ||
BC005480.1 | - | 8 - 9 | 977 | 69 | ||
BC007145.1 | - | 19 - 20 | 2875 | 3490671 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |