Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163626.1 | NP_001157098.1 | 5 - 6 | 893 | 68 | |
NM_172204.4 | NP_757341.3 | 5 - 6 | 893 | 68 | ||
GenBank mRNA | AF539798.1 | - | 5 - 6 | 612 | 68 | |
AK087263.1 | - | 5 - 6 | 819 | 68 | ||
AY255770.1 | - | 5 - 6 | 612 | 68 | ||
BC047532.1 | - | 5 - 6 | 884 | 4988389 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |