Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172307.3 | NP_758511.1 | 9 - 10 | 1395 | 89 | |
GenBank mRNA | AK077220.1 | - | 1341 | 89 | ||
AK148325.1 | - | 9 - 10 | 1360 | 89 | ||
AK164486.1 | - | 9 - 10 | 1331 | 89 | ||
BC038343.1 | - | 9 - 10 | 1322 | 4460515 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |