Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019509.mm.1 |
NM_172406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019509.mm.1 |
NM_172406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019509.mm.1 |
TC0100002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019508.mm.1 |
NM_172406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019508.mm.1 |
NM_172406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019508.mm.1 |
TC0100002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019512.mm.1 |
NM_172406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019512.mm.1 |
NM_172406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019512.mm.1 |
TC0100002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019510.mm.1 |
NM_172406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019510.mm.1 |
NM_172406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019510.mm.1 |
TC0100002392.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019507.mm.1 |
NM_172406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019507.mm.1 |
NM_172406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019507.mm.1 |
TC0100002392.mm.1 |
MTA TC |