Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172443.3 | NP_766031.2 | 6 - 7 | 1416 | 58 | |
GenBank mRNA | AK047555.1 | - | 6 - 7 | 1417 | 58 | |
BC057634.1 | - | 6 - 7 | 1375 | 2866041 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |