Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172444.4 | NP_766032.1 | 1 - 2 | 240 | 77 | |
GenBank mRNA | AK045414.1 | - | 1 - 2 | 156 | 77 | |
BC139329.1 | - | 1 - 2 | 133 | 8862351 | 77 | |
CV559893.1 | - | 1 - 2 | 240 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |