Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205367.1 | NP_001192296.1 | 3 - 4 | 467 | 90 | |
NM_009859.4 | NP_033989.2 | 4 - 5 | 470 | 90 | ||
GenBank mRNA | AK088997.1 | - | 4 - 5 | 257 | 90 | |
AK144127.1 | - | 4 - 5 | 259 | 90 | ||
AK147986.1 | - | 4 - 5 | 262 | 90 | ||
AK220447.1 | - | 3 - 4 | 240 | 90 | ||
BC058587.1 | - | 3 - 4 | 271 | 6529169 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |