Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011023.3 | NP_035153.1 | 2 - 3 | 173 | 77 | |
GenBank mRNA | AK042975.1 | - | 2 - 3 | 137 | 77 | |
AK050758.1 | - | 2 - 3 | 134 | 77 | ||
AK083434.1 | - | 2 - 3 | 135 | 77 | ||
AK083671.1 | - | 2 - 3 | 117 | 77 | ||
BC057105.1 | - | 1 - 2 | 83 | 6416436 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |