Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011480.3 | NP_035610.1 | 1 - 2 | 158 | 62 | |
GenBank mRNA | AK052628.1 | - | 1 - 2 | 159 | 62 | |
AK150052.1 | - | 1 - 2 | 147 | 62 | ||
AK154424.1 | - | 1 - 2 | 141 | 62 | ||
AK169607.1 | - | 1 - 2 | 159 | 62 | ||
BC056922.1 | - | 1 - 2 | 140 | 6824948 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Estrogen Panel 1700 Inventoried Probe spans exons |