Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011536.2 | NP_035666.2 | 3 - 4 | 418 | 84 | |
NM_172798.1 | NP_766386.1 | 2 - 3 | 368 | 84 | ||
GenBank mRNA | AB221665.1 | - | 3 - 4 | 418 | 84 | |
AK028407.1 | - | 2 - 3 | 302 | 84 | ||
AK139949.1 | - | 2 - 3 | 340 | 84 | ||
AK147984.1 | - | 2 - 3 | 286 | 84 | ||
AY075134.1 | - | 3 - 4 | 418 | 84 | ||
BQ560189.1 | - | 2 - 3 | 368 | 84 | ||
U57329.1 | - | 2 - 3 | 145 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |