Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013768.3 | NP_038796.2 | 1 - 2 | 223 | 69 | |
GenBank mRNA | AF167573.1 | - | 1 - 2 | 91 | 69 | |
AK076346.1 | - | 1 - 2 | 114 | 69 | ||
AK084106.1 | - | 1 - 2 | 224 | 69 | ||
AK132414.1 | - | 1 - 2 | 115 | 69 | ||
AK165165.1 | - | 1 - 2 | 141 | 69 | ||
BC023905.1 | - | 1 - 2 | 170 | 5321370 | 69 | |
BY064945.1 | - | 1 - 2 | 279 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |