Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF316872.1 | - | 3 - 4 | 823 | 63 | |
AK215426.1 | - | 3 - 4 | 264 | 63 | ||
AK220318.1 | - | 3 - 4 | 799 | 63 | ||
BC018561.1 | - | 3 - 4 | 671 | 63 | ||
BC018594.1 | - | 3 - 4 | 808 | 63 | ||
BC069968.1 | - | 4 - 5 | 703 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |