Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 8 - 9 | 1356 | 62 | |
GenBank mRNA | AK016437.1 | - | 6 - 7 | 715 | 62 | |
AK039189.1 | - | 8 - 9 | 1340 | 62 | ||
AK040513.1 | - | 7 - 8 | 1086 | 62 | ||
BC020351.1 | - | 8 - 9 | 1569 | 62 | ||
BC032150.1 | - | 4 - 5 | 562 | 5004308 | 62 | |
BC076571.1 | - | 8 - 9 | 1253 | 30356163 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |